Learn More
Electron Microscopy Sciences LifterSlip™ Premium Printed Coverglass, 25 x 44 mm 1 OZ
SDP
Catalog No. 501899774
Encompass_Preferred
LifterSlip offers a simple solution to the problem of hybridization non-uniformity. An innovative raised edge design provides separation and allows for even dispersal of hybridization solution between the DNA chip and coverslip. LifterSlips allow for increased data quality by eliminating gradients caused by floating standard coverslips on hybridization solutions. The printed bars allow for easy injection of hybridization solution under the pre-positioned coverslips. Capillary attraction ensures that LifterSlip stays in place.
LifterSlip is #2 thickness (0.20-0.22 mm) and the bar height is 0.04-0.06 mm.
Size: 25 x 44 mm
Catalog No. 50-189-9774 Supplier Electron Microscopy Sciences Supplier No. 7218644
LifterSlip offers a simple solution to the problem of hybridization non-uniformity. An innovative raised edge design provides separation and allows for even dispersal of hybridization solution between the DNA chip and coverslip. LifterSlips allow for increased data quality by eliminating gradients caused by floating standard coverslips on hybridization solutions. The printed bars allow for easy injection of hybridization solution under the pre-positioned coverslips. Capillary attraction ensures that LifterSlip stays in place.
LifterSlip is #2 thickness (0.20-0.22 mm) and the bar height is 0.04-0.06 mm.
Size: 25 x 44 mm