Learn More
Innovative Research Inc Cow Dxyrbnclc Acd Cellulose 2S
Catalog No. 502744627
Encompass_Preferred
Bovine Deoxyribonucleic Acid Cellulose Double Strand 1gm - Bovine Deoxyribonucleic Acid Cellulose Double Strand from Innovative Research is prepared by a native, double-stranded calf thymus DNA being covalently bound to cellulose. One gram of DNA-cellulose does swell to 3 - 4 ml when fully hydrated. This is a dry powder. This product is useful for the purification of many DNA binding proteins such as polymerases, transcription factors, and terminators, and the like. Calf thymus DNA Cellulose, Double-Stranded.
Catalog No. 50-274-4627 Supplier Innovative Research Inc Supplier No. IBODNACSO2S1GM
Bovine Deoxyribonucleic Acid Cellulose Double Strand 1gm - Bovine Deoxyribonucleic Acid Cellulose Double Strand from Innovative Research is prepared by a native, double-stranded calf thymus DNA being covalently bound to cellulose. One gram of DNA-cellulose does swell to 3 - 4 ml when fully hydrated. This is a dry powder. This product is useful for the purification of many DNA binding proteins such as polymerases, transcription factors, and terminators, and the like. Calf thymus DNA Cellulose, Double-Stranded.