Learn More
Innovative Research Inc Cow Myoglobin Pur Lyo 250mg
Catalog No. 502744623
Encompass_Preferred
Bovine Myoglobin Purified Lyophilized 250mg - Bovine Myoglobin Purified Lyophilized from Innovative Research is a dialyzed, lyophilized powder. A core responsibility of this globular protein is to store oxygen in cardiac and skeletal muscle. This product should be protected from moisture.
Catalog No. 50-274-4623 Supplier Innovative Research Inc Supplier No. IBOMBLY250MG
Bovine Myoglobin Purified Lyophilized 250mg - Bovine Myoglobin Purified Lyophilized from Innovative Research is a dialyzed, lyophilized powder. A core responsibility of this globular protein is to store oxygen in cardiac and skeletal muscle. This product should be protected from moisture.