Learn More
Innovative Research Inc Cow RBCs Packed 100 1L
Catalog No. 502744619
Encompass_Preferred
Bovine Red Blood Cells Packed 100% 1000ml - Bovine Red Blood Cells from Innovative Research are manufactured using whole blood collections from cows located in the United States. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. The red blood cells are supplied as a 100% suspension in phosphate buffered saline (PBS). Red blood cells are perishable and are collected and processed upon receipt of order.
Catalog No. 50-274-4619 Supplier Innovative Research Inc Supplier No. IBORBC100P1000ML
Bovine Red Blood Cells Packed 100% 1000ml - Bovine Red Blood Cells from Innovative Research are manufactured using whole blood collections from cows located in the United States. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. The red blood cells are supplied as a 100% suspension in phosphate buffered saline (PBS). Red blood cells are perishable and are collected and processed upon receipt of order.