Learn More
Synthware™ Cow Receiver
GREENER_CHOICESDP
Catalog No. 31501050 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint:
14/20
19/22
24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint
31-501-050 24/40
31-501-044 14/20
31-501-046 19/22
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-501-050 Supplier Synthware™ Supplier No. D202424
Add to Cart
Add to Cart
For use with distribution adapter/receiving set
Specifications
For Use With (Application) Organic Synthesis
Type Cow Type Distillation Receiver
Material Borosilicate Glass
For Use With (Equipment) Distribution adapter
Quantity 1 Ea.
Standard Taper Joint 24/40