Learn More
Sigma Aldrich Fine Chemicals Biosciences COware gas reactor reactio
Supplier:  Sigma Aldrich Fine Chemicals Biosciences STW51EA
Encompass
Two-chamber glass apparatus are ideal for reaction scales of0.1-1 mmol (total volume 20 mL)1.0-5.0 mmol (total volume 100 mL)5.0-20.0 mmol (total volume 400 mL)Each unit includes two screw caps two stabilizing PTfE discs (recyclable) ten single use silicone/PTfE septa (in combination with the stabilizing discs each septum allows for a maximum of 2 piercings using 21G/A˜0.80 mm syringes or smaller).Order replacement PTfE/silicone septa STW3.
Catalog No. 50-290-6574