Learn More
Medchemexpress LLC COX-1-IN-3 | 20170-32-5 | 99.9% | 278.39 | 50 G
SDP
Catalog No. 5000366962
Encompass_Preferred
COX-1-IN-3 (compound 19) is a COX-1 inhibitor with non-steroidal anti-inflammatory activity. It is intended for research use only.
  • COX-1 inhibitor
  • Non-steroidal anti-inflammatory activity
  • High purity (99.95%)
  • Available in various quantities and solution formats
Catalog No. 50-003-66962 Supplier Medchemexpress LLC Supplier No. HYW00934750G
COX-1-IN-3 (compound 19) is a COX-1 inhibitor with non-steroidal anti-inflammatory activity. It is intended for research use only.
  • COX-1 inhibitor
  • Non-steroidal anti-inflammatory activity
  • High purity (99.95%)
  • Available in various quantities and solution formats