Learn More
Cayman Chemical COX-1 (human, recombinant), Purity: ≥85% estimated by SDS-PAGE, 5 kU
SDP
Catalog No. 502967428
Encompass_Preferred
Source: Active recombinant human His-tagged COX-1 expressed in insect cells • Amino acids: 24-599 • MW: ~70 kDa
Catalog No. 50-296-7428 Supplier Cayman Chemical Supplier No. 176165 KU
Source: Active recombinant human His-tagged COX-1 expressed in insect cells • Amino acids: 24-599 • MW: ~70 kDa