Learn More
Enzo Life Sciences COX-2 (human), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900094
Encompass_Preferred
Most sensitive COX-II immunoassay kit commercially available for inflammation research. The COX-2 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 2 hours. Alternative Name: Cyclooxygenase-2. Prostaglandin H synthase 2. Sensitivity: 0.25 ng/ml (range 1.09 - 70 ng/ml). Assay Time: 2 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Human. Contents: Microtiter plate, Labeled antibody concentrate, Antibody diluent, Assay buffer, Wash buffer concentrate, Standard, TMB Substrate, Stop solution
Catalog No. 50-658-139