Learn More
TARGETMOL CHEMICALS INC COX-2-IN-1 5MG
SDP
Catalog No. 502581771
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. COX-2-IN-1 is a potent and selective COX-2 inhibitor (IC50 3.9 uM). purity: 100%
Catalog No. 50-258-1771 Supplier TARGETMOL CHEMICALS INC Supplier No. T100335MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. COX-2-IN-1 is a potent and selective COX-2 inhibitor (IC50 3.9 uM). purity: 100%