Learn More
AdipoGen COX-2 Inhibitor 8c (1 mg)
SDP
Supplier:  AdipoGen AGCR13606M001
Encompass_Preferred
Chemical. CAS: 1259295-44-7. Formula: C12H11NO2S2. MW: 265.4. Potent cyclooxygenase-2 (COX-2) inhibitor. Inhibits inflammatory prostaglandin E2 (PGE2) production.
Catalog No. 50-205-3975