Learn More
Cayman Chemical COX Activity Assay Kit, 96 Well
SDP
Catalog No. 502893874
Encompass_Preferred
Cayman’s COX Activity Assay Kit measures the peroxidase activity of COX. The peroxidase activity is assayed colorimetrically by monitoring the appearance of oxidized N,N,N’,N’-tetramethyl-p-phenylenediamine (TMPD) at 590 nm.{8581} It can be used with tissue homogenates and purified enzyme preparations. The kit includes isozyme-specific inhibitors for distinguishing COX-2 activity from COX-1 activity.
Catalog No. 50-289-3874 Supplier Cayman Chemical Supplier No. 76015196 WELL
Cayman’s COX Activity Assay Kit measures the peroxidase activity of COX. The peroxidase activity is assayed colorimetrically by monitoring the appearance of oxidized N,N,N’,N’-tetramethyl-p-phenylenediamine (TMPD) at 590 nm.{8581} It can be used with tissue homogenates and purified enzyme preparations. The kit includes isozyme-specific inhibitors for distinguishing COX-2 activity from COX-1 activity.