Learn More
Aviva Systems Biology Corp COX15 Antibody - N-terminal region (ARP46442_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46442_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against COX15. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human COX15. Alternate names: -ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-59327