Learn More
Enzo Life Sciences COX-2 monoclonal antibody (AS66) (100µg)
SDP
Supplier:  Enzo Life Sciences ADICOX030E
Encompass_Preferred
Clone: AS66. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human COX-2 (cyclooxygenase-2). UniProt ID: P35354. Species reactivity: Human. Creossreactivity: Zero percent cross-reactivity to human recombinant COX-1 by ELISA. Applications: ELISA, Flow Cytometry, WB. Recommended Dilutions: Western Blot (1-5 µg/ml) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Purified. Formulation: Liquid. In PBS containing 0.05% sodium azide.
Catalog No. 50-200-7182