Learn More
Sino Biological COX-2/PTGS2 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 106147T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse COX-2/PTGS2, and purified by antigen affinity chromatography.
Catalog No. 50-224-8290