Learn More
Aviva Systems Biology Corp COX4I1 Antibody - N-terminal region (ARP42784_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42784_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against COX4I1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human COX4I1. Alternate names: COX4, COXIV, MGC72016, COX4-1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58332