Learn More
Aviva Systems Biology Corp Cox6a1 Antibody - C-terminal region (ARP41557_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41557_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cox6a1. It was validated on Western Blot. . Alternate names: VIaLApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-284-941