Learn More
Aviva Systems Biology Corp COX6A1 Antibody - N-terminal region (ARP46438_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46438_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against COX6A1. It was validated on Western Blot. . Alternate names: COX6A, COX6AL, MGC104500ApplicationWB. Species reactivity: Human
Catalog No. 50-285-258