Learn More
Aviva Systems Biology Corp COX7B Antibody - N-terminal region (ARP51975_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51975_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against COX7B. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human COX7B. Alternate names: -ApplicationWB. Species reactivity: Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-618