Learn More
Cayman Chemical Coydals duls max. Coydn 10mg
SDP
Catalog No. 502910668
Encompass_Preferred
An aporphine alkaloid with diverse biological activities; inhibits the proliferation murine L1210 lymphocytic leukemia, B16/F10 melanoma, and P388 leukemia cells from 10-100 UG/ml; binds to μ-opioid receptors (Ki = 2.82 µM) and induces [35S]GTPγS binding in CHO cells expressing human μ-opioid receptors (EC50 = 0.51 µM); active against L. donovani promastigotes (IC50 = 26.7 µM)
Catalog No. 50-291-0668 Supplier Cayman Chemical Supplier No. 3502810 MG
An aporphine alkaloid with diverse biological activities; inhibits the proliferation murine L1210 lymphocytic leukemia, B16/F10 melanoma, and P388 leukemia cells from 10-100 UG/ml; binds to μ-opioid receptors (Ki = 2.82 µM) and induces [35S]GTPγS binding in CHO cells expressing human μ-opioid receptors (EC50 = 0.51 µM); active against L. donovani promastigotes (IC50 = 26.7 µM)