Learn More
Cayman Chemical Coydals ynusuo Coydaln 100mg
SDP
Catalog No. 502940706
Encompass_Preferred
An alkaloid with diverse biological activities; inhibits AChE (IC50 = 15 μM); nematocidal against S. ratti and S. venezuelensis third instar larvae (PC50s = 18 and 30 μM, respectively); inhibits thrombin-induced platelet aggregation in vitro (IC50 = 54.16 μg/ml); increases gastric emptying in rat models of apomorphine- and laparotomy-induced delayed gastric emptying at 1 and 3 mg/kg
Catalog No. 50-294-0706 Supplier Cayman Chemical Supplier No. 27654100 MG
An alkaloid with diverse biological activities; inhibits AChE (IC50 = 15 μM); nematocidal against S. ratti and S. venezuelensis third instar larvae (PC50s = 18 and 30 μM, respectively); inhibits thrombin-induced platelet aggregation in vitro (IC50 = 54.16 μg/ml); increases gastric emptying in rat models of apomorphine- and laparotomy-induced delayed gastric emptying at 1 and 3 mg/kg