Learn More
Cayman Chemical Corydaline, Purity: ≥98%, CAS: 518-69-4, 50 mg
SDP
Catalog No. 502940853
Encompass_Preferred
An alkaloid with diverse biological activities; inhibits AChE (IC50 = 15 μM); nematocidal against S. ratti and S. venezuelensis third instar larvae (PC50s = 18 and 30 μM, respectively); inhibits thrombin-induced platelet aggregation in vitro (IC50 = 54.16 μg/ml); increases gastric emptying in rat models of apomorphine- and laparotomy-induced delayed gastric emptying at 1 and 3 mg/kg
Catalog No. 50-294-0853 Supplier Cayman Chemical Supplier No. 2765450 MG
An alkaloid with diverse biological activities; inhibits AChE (IC50 = 15 μM); nematocidal against S. ratti and S. venezuelensis third instar larvae (PC50s = 18 and 30 μM, respectively); inhibits thrombin-induced platelet aggregation in vitro (IC50 = 54.16 μg/ml); increases gastric emptying in rat models of apomorphine- and laparotomy-induced delayed gastric emptying at 1 and 3 mg/kg