Learn More
Cayman Chemical CP 100,356 (hydrochloride), Purity: ≥98%, CAS: 142715-48-8, 10 mg
SDP
Catalog No. 502950346
Encompass_Preferred
A P-glycoprotein inhibitor (IC50 = 0.5 µM); selective for P-gp over MRP2 at 15 µM, CYP1A2, CYP2C9, CYP2C19, CYP2D6, and CYP3A4 (IC50s = >50 µM for all), and OATP2B1 (IC50 = ~66 µM) but does inhibit BCRP (IC50 = 1.5 µM); increases the rate of prazosin clearance in rats at 24 mg/kg when administered in combination with prazosin
Catalog No. 50-295-0346 Supplier Cayman Chemical Supplier No. 3285310 MG
A P-glycoprotein inhibitor (IC50 = 0.5 µM); selective for P-gp over MRP2 at 15 µM, CYP1A2, CYP2C9, CYP2C19, CYP2D6, and CYP3A4 (IC50s = >50 µM for all), and OATP2B1 (IC50 = ~66 µM) but does inhibit BCRP (IC50 = 1.5 µM); increases the rate of prazosin clearance in rats at 24 mg/kg when administered in combination with prazosin