Learn More
Medchemexpress LLC Cp-100356 monohydrochloride | 142715-48-8 | MFCD00911858 | >98.0% | 597.10 g·mol⁻¹ | C31H37ClN4O6 | 1 ML
SDP
Catalog No. 5000243345
Encompass_Preferred
CP-100356 hydrochloride is a research-grade inhibitor of multidrug transporters used to study transporter-mediated drug disposition and drug-drug interactions. It is supplied in both solid form and as a ready-to-use 10 mM solution in DMSO for in vitro assays, and it is commonly used in pharmacokinetic and cellular uptake studies.
  • Potent inhibitor of P-gp and BCRP transporters.
  • Available as a 10 mM solution in DMSO or as solid packs (5-100 mg).
  • High purity suitable for in vitro assays (>98% by HPLC).
  • Well characterized molecular properties for experimental reproducibility.
  • Used for transporter assays and drug-drug interaction studies.
Catalog No. 50-002-43345 Supplier Medchemexpress LLC Supplier No. HY10834710MM 1ML
CP-100356 hydrochloride is a research-grade inhibitor of multidrug transporters used to study transporter-mediated drug disposition and drug-drug interactions. It is supplied in both solid form and as a ready-to-use 10 mM solution in DMSO for in vitro assays, and it is commonly used in pharmacokinetic and cellular uptake studies.
  • Potent inhibitor of P-gp and BCRP transporters.
  • Available as a 10 mM solution in DMSO or as solid packs (5-100 mg).
  • High purity suitable for in vitro assays (>98% by HPLC).
  • Well characterized molecular properties for experimental reproducibility.
  • Used for transporter assays and drug-drug interaction studies.