Learn More
Cayman Chemical CP 154 526hydrochlorIde 10mg
SDP
Catalog No. 502982740
Encompass_Preferred
An antagonist of CRF1 (Ki = 2.7 nM); selective for CRF1 over CRF2 (Ki = >10 μM); inhibits CRF-induced increases in ACTH levels in rat plasma (ID50 = 10 mg/kg); increases the time spent in the open arms of the elevated plus maze in rats at 1 mg/kg; decreases immobility time in the forced swim test in a rat model of depressive-like behavior at 10 mg/kg, twice per day for 14 days; decreases stress-induced reinstatement of cocaine or heroin self-administration in cocaine- and heroin-trained rats, respectively, at 15 and 30 mg/kg
Catalog No. 50-298-2740 Supplier Cayman Chemical Supplier No. 2073210 MG
An antagonist of CRF1 (Ki = 2.7 nM); selective for CRF1 over CRF2 (Ki = >10 μM); inhibits CRF-induced increases in ACTH levels in rat plasma (ID50 = 10 mg/kg); increases the time spent in the open arms of the elevated plus maze in rats at 1 mg/kg; decreases immobility time in the forced swim test in a rat model of depressive-like behavior at 10 mg/kg, twice per day for 14 days; decreases stress-induced reinstatement of cocaine or heroin self-administration in cocaine- and heroin-trained rats, respectively, at 15 and 30 mg/kg