Learn More
Abcam CP 154526 hydrochloride, non-peptide CRF1 receptor antagonist, 10MG
Catalog No. 502706624
Encompass_Preferred
MW 400.99 g/mol, Purity >99%. Potent, selective, non-peptide corticotropin factor receptor-1 antagonist (Ki = 10 nM). Blood-brain barrier permeable. Neuroprotective and antistress agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6624 Supplier Abcam Supplier No. AB14142910MG
MW 400.99 g/mol, Purity >99%. Potent, selective, non-peptide corticotropin factor receptor-1 antagonist (Ki = 10 nM). Blood-brain barrier permeable. Neuroprotective and antistress agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement