Learn More
Cayman Chemical CP 24,879 (hydrochloride), Purity: ≥98%, CAS: 10141-51-2, 25 mg
SDP
Catalog No. 502891402
Encompass_Preferred
A Δ5/Δ6 desaturase inhibitor; reduces liver arachidonate content by ~50% in mice at 3 mg/kg; increases mead acid content and inhibits leukotriene C4 synthesis in murine mastocytoma cells at 5 µM
Catalog No. 50-289-1402 Supplier Cayman Chemical Supplier No. 1012025 MG
A Δ5/Δ6 desaturase inhibitor; reduces liver arachidonate content by ~50% in mice at 3 mg/kg; increases mead acid content and inhibits leukotriene C4 synthesis in murine mastocytoma cells at 5 µM