Learn More
Medchemexpress LLC CP-24879 hydrochlori 10mM 1mL | 10141-51-2 | 1 ML
SDP
Catalog No. 5000226897
Encompass_Preferred
CP-24879 hydrochloride is a research compound that potently and selectively inhibits delta5 and delta6 desaturases. It is supplied as a 10 mM solution in DMSO (1 mL) for biochemical and cell-based studies of lipid metabolism and inflammatory pathways. CAS 10141-51-2; molecular formula C11H18ClNO; molecular weight 215.72 g·mol⁻¹; reported purity 99.59%.
  • Potent dual delta5/delta6 desaturase inhibitor.
  • Supplied as a 10 mM solution in DMSO, 1 mL format.
  • High reported purity suitable for research use.
  • Appropriate for biochemical and cell-based lipid metabolism studies.
  • Molecular formula C11H18ClNO; molecular weight 215.72 g·mol⁻¹.
Catalog No. 50-002-26897 Supplier Medchemexpress LLC Supplier No. HY11531910MM 1ML
CP-24879 hydrochloride is a research compound that potently and selectively inhibits delta5 and delta6 desaturases. It is supplied as a 10 mM solution in DMSO (1 mL) for biochemical and cell-based studies of lipid metabolism and inflammatory pathways. CAS 10141-51-2; molecular formula C11H18ClNO; molecular weight 215.72 g·mol⁻¹; reported purity 99.59%.
  • Potent dual delta5/delta6 desaturase inhibitor.
  • Supplied as a 10 mM solution in DMSO, 1 mL format.
  • High reported purity suitable for research use.
  • Appropriate for biochemical and cell-based lipid metabolism studies.
  • Molecular formula C11H18ClNO; molecular weight 215.72 g·mol⁻¹.