Learn More
Medchemexpress LLC CP-24879 hydrochlori 1mg | 10141-51-2 | 215.72 | 1 MG
SDP
Catalog No. 5000229595
Encompass_Preferred
CP-24879 hydrochloride is a small-molecule research reagent that functions as a potent and selective combined delta5-desaturase/delta6-desaturase (Δ5/Δ6) inhibitor. It reduces intracellular lipid accumulation and inflammatory injury in hepatocyte models and is used in studies of fatty acid metabolism and non-alcoholic steatohepatitis. This compound is supplied for laboratory research use only.
  • Potent combined Δ5/Δ6 desaturase inhibition
  • Applied in lipid metabolism and NASH research
  • High purity (99.59% by LCMS)
  • Solid, off-white to light brown appearance
  • Available in small research quantities (for example, 1 mg)
Catalog No. 50-002-29595 Supplier Medchemexpress LLC Supplier No. HY1153191MG
CP-24879 hydrochloride is a small-molecule research reagent that functions as a potent and selective combined delta5-desaturase/delta6-desaturase (Δ5/Δ6) inhibitor. It reduces intracellular lipid accumulation and inflammatory injury in hepatocyte models and is used in studies of fatty acid metabolism and non-alcoholic steatohepatitis. This compound is supplied for laboratory research use only.
  • Potent combined Δ5/Δ6 desaturase inhibition
  • Applied in lipid metabolism and NASH research
  • High purity (99.59% by LCMS)
  • Solid, off-white to light brown appearance
  • Available in small research quantities (for example, 1 mg)