Learn More
Medchemexpress LLC CP-289,503 | 1349637-14-4 | 98.2% | 491.06 | 5 MG
SDP
Catalog No. 5000393020
Encompass_Preferred
CP-289,503 is an inhibitor of the complement C5a receptor with an IC50 of 1 μM. It blocks C5a signaling and can inhibit a variety of inflammatory diseases by preventing C5a from stimulating the upregulation of cell surface integrins and degranulation of inflammatory cells.
  • Inhibitor of the complement C5a receptor
  • Blocks C5a signaling
  • Suitable for research into inflammatory and autoimmune diseases
Catalog No. 50-003-93020 Supplier Medchemexpress LLC Supplier No. HY1609015MG
CP-289,503 is an inhibitor of the complement C5a receptor with an IC50 of 1 μM. It blocks C5a signaling and can inhibit a variety of inflammatory diseases by preventing C5a from stimulating the upregulation of cell surface integrins and degranulation of inflammatory cells.
  • Inhibitor of the complement C5a receptor
  • Blocks C5a signaling
  • Suitable for research into inflammatory and autoimmune diseases