Learn More
Medchemexpress LLC CP-289 503 | 1349637-14-4 | 98.2% | 491.06 | 50 MG
SDP
Catalog No. 5000393068
Encompass_Preferred
CP-289,503 is an inhibitor of the complement C5a receptor with an IC50 of 1 μM. C5a acts as an activator of leukocytes and phagocytes during complement system activation. The C5a receptor can bind to C5a, which can stimulate the upregulation of cell surface integrins and degranulation of inflammatory cells, leading to endothelial cell damage. C5a receptor inhibitors can block C5a signaling and inhibit a variety of inflammatory diseases.
  • Inhibitor of the complement C5a receptor.
  • Blocks C5a signaling.
  • Inhibits a variety of inflammatory diseases.
Catalog No. 50-003-93068 Supplier Medchemexpress LLC Supplier No. HY16090150MG
CP-289,503 is an inhibitor of the complement C5a receptor with an IC50 of 1 μM. C5a acts as an activator of leukocytes and phagocytes during complement system activation. The C5a receptor can bind to C5a, which can stimulate the upregulation of cell surface integrins and degranulation of inflammatory cells, leading to endothelial cell damage. C5a receptor inhibitors can block C5a signaling and inhibit a variety of inflammatory diseases.
  • Inhibitor of the complement C5a receptor.
  • Blocks C5a signaling.
  • Inhibits a variety of inflammatory diseases.