Learn More
Apexbio Technology LLC CP 339818 hydrochloride 10mg
SDP
Catalog No. 5000088575
Encompass_Preferred
CP 339818 hydrochloride (CAS 478341-55-8) is a small-molecule antagonist targeting the Kv1 3 potassium channel It is designed to selectively inhibit Kv1 3-mediated ion conductance thereby modulating T-cell activation CP 339818 hydrochloride exerts its biological activity primarily through stabilization of the Kv1 3 channel s C-type inactivation state In in vitro studies CP 339818 hydrochloride demonstrates potent inhibition of Kv1 3 channels with an IC50 value of approximately 200 nM Based on these pharmacological properties CP 339818 hydrochloride holds research potential in the investigation of immune modulation mechanisms relevant to autoimmune and inflammatory disorders
Catalog No. 50-000-88575 Supplier Apexbio Technology LLC Supplier No. B672110
CP 339818 hydrochloride (CAS 478341-55-8) is a small-molecule antagonist targeting the Kv1 3 potassium channel It is designed to selectively inhibit Kv1 3-mediated ion conductance thereby modulating T-cell activation CP 339818 hydrochloride exerts its biological activity primarily through stabilization of the Kv1 3 channel s C-type inactivation state In in vitro studies CP 339818 hydrochloride demonstrates potent inhibition of Kv1 3 channels with an IC50 value of approximately 200 nM Based on these pharmacological properties CP 339818 hydrochloride holds research potential in the investigation of immune modulation mechanisms relevant to autoimmune and inflammatory disorders