Learn More
Medchemexpress LLC CP 339818 (hydrochloride) | 478341-55-8 | 99.6% | 340.89 | 25 MG
SDP
Catalog No. 5000347736
Encompass_Preferred
CP 339818 hydrochloride is a non-peptide blocker for Kv1.3 and Kv1.4 channels. It also inhibits HCN channels and exhibits significantly weaker blocking effects on several other Kv channels. This compound selectively blocks Kv1.3, thereby inhibiting the activation process of human T cells, and is useful for studying the physiological functions of HCN and Kv channels.
  • Kv1.3 channel blocker (IC50 = 200 nM).
  • Kv1.4 channel blocker.
  • Inhibits HCN channels (HCN1 and HCN4).
  • Selectively inhibits human T cell activation.
  • Suitable for studying physiological functions of HCN and Kv channels.
Catalog No. 50-003-47736 Supplier Medchemexpress LLC Supplier No. 25MGHY108574
CP 339818 hydrochloride is a non-peptide blocker for Kv1.3 and Kv1.4 channels. It also inhibits HCN channels and exhibits significantly weaker blocking effects on several other Kv channels. This compound selectively blocks Kv1.3, thereby inhibiting the activation process of human T cells, and is useful for studying the physiological functions of HCN and Kv channels.
  • Kv1.3 channel blocker (IC50 = 200 nM).
  • Kv1.4 channel blocker.
  • Inhibits HCN channels (HCN1 and HCN4).
  • Selectively inhibits human T cell activation.
  • Suitable for studying physiological functions of HCN and Kv channels.