Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CP 339818 hydrochloride | 478341-55-8 | 99.6% | 340.89 g/mol | C21H25ClN2 | 5 MG
SDP

Catalog No. 5000264141
Encompass_Preferred

CP 339818 hydrochloride is a small-molecule, non-peptide ion-channel inhibitor used in electrophysiology and pharmacology research. It blocks Kv1.3 and Kv1.4 channels and inhibits HCN channels, making it useful for studying Kv- and HCN-mediated currents and T-cell activation.

  • Non-peptide Kv1.3 and Kv1.4 channel blocker.
  • Inhibits HCN channel isoforms with micromolar potency.
  • High-purity white to off-white solid suitable for research use.
  • Soluble in DMSO up to 62.5 mg/mL.
  • Store sealed at 4°C; in solution, keep at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Molecular weight 340.89 g/mol; formula C21H25ClN2.

Catalog No. 50-002-64141 Supplier Medchemexpress LLC Supplier No. HY1085745MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CP 339818 hydrochloride is a small-molecule, non-peptide ion-channel inhibitor used in electrophysiology and pharmacology research. It blocks Kv1.3 and Kv1.4 channels and inhibits HCN channels, making it useful for studying Kv- and HCN-mediated currents and T-cell activation.

  • Non-peptide Kv1.3 and Kv1.4 channel blocker.
  • Inhibits HCN channel isoforms with micromolar potency.
  • High-purity white to off-white solid suitable for research use.
  • Soluble in DMSO up to 62.5 mg/mL.
  • Store sealed at 4°C; in solution, keep at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Molecular weight 340.89 g/mol; formula C21H25ClN2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.