Learn More
Medchemexpress LLC HY-14130 5mg Medchemexpress, CP 376395 CAS:175140-00-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY141305MG
Encompass_Preferred
Medchemexpress, HY-14130 5mg CP 376395 CAS:175140-00-8 CP 376395 is a potent and selective Corticotropin releasing factor 1 (CRF1) receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1563