Learn More
Medchemexpress LLC Cp-409092 hydrochloride | 225240-86-8 | 99.8% | 333.81 g/mol | C17H20ClN3O2 | 10 MG
SDP
Catalog No. 5000249039
Encompass_Preferred
CP-409092 hydrochloride is a research-use small molecule described as a partial agonist of GABAA receptors with reported anti-anxiety activity. It is supplied as a solid for laboratory research and is not intended for human therapeutic use.
  • Partial agonist of GABAA receptor, with anti-anxiety activity.
  • High purity: 99.8% (reported).
  • Solid, white to off-white appearance for handling and formulation.
  • Recommended storage: 4°C sealed; in solvent, -80°C (up to 6 months) or -20°C (up to 1 month).
  • Available in small research pack sizes; common pack: 10 MG.
  • For research use only; not for clinical or human use.
Catalog No. 50-002-49039 Supplier Medchemexpress LLC Supplier No. HY101639A10MG
CP-409092 hydrochloride is a research-use small molecule described as a partial agonist of GABAA receptors with reported anti-anxiety activity. It is supplied as a solid for laboratory research and is not intended for human therapeutic use.
  • Partial agonist of GABAA receptor, with anti-anxiety activity.
  • High purity: 99.8% (reported).
  • Solid, white to off-white appearance for handling and formulation.
  • Recommended storage: 4°C sealed; in solvent, -80°C (up to 6 months) or -20°C (up to 1 month).
  • Available in small research pack sizes; common pack: 10 MG.
  • For research use only; not for clinical or human use.