Learn More
Medchemexpress LLC Cp-409092 hydrochloride | 225240-86-8 | 99.8% | 333.81 g/mol | C17H20ClN3O2 | 25 MG
SDP
Catalog No. 5000245445
Encompass_Preferred
CP-409092 hydrochloride is a research chemical that functions as a partial agonist of the GABAA receptor and has reported anxiolytic activity. Supplied as the hydrochloride salt, it is provided as a high-purity solid for in vitro and in vivo pharmacology studies.
  • Acts as a partial agonist of the GABAA receptor.
  • Demonstrates reported anti-anxiety (anxiolytic) activity in pharmacological assays.
  • High purity (≈99.8%) with white to off-white solid appearance.
  • Soluble in water and DMSO with ultrasonic assistance for stock preparation.
  • Documented with COA, SDS, and data sheet for research handling.
Catalog No. 50-002-45445 Supplier Medchemexpress LLC Supplier No. HY101639A25MG
CP-409092 hydrochloride is a research chemical that functions as a partial agonist of the GABAA receptor and has reported anxiolytic activity. Supplied as the hydrochloride salt, it is provided as a high-purity solid for in vitro and in vivo pharmacology studies.
  • Acts as a partial agonist of the GABAA receptor.
  • Demonstrates reported anti-anxiety (anxiolytic) activity in pharmacological assays.
  • High purity (≈99.8%) with white to off-white solid appearance.
  • Soluble in water and DMSO with ultrasonic assistance for stock preparation.
  • Documented with COA, SDS, and data sheet for research handling.