Learn More
TARGETMOL CHEMICALS INC CP 461 5MG
SDP
Catalog No. 502580383
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. CP 461 (UNII-68OJX9I7DT) a specific PDE2A inhibitor is a novel pro-apoptotic compound that inhibits cyclic GMP phosphodiesterase but not cyclooxygenase-1 or -2. CP 461 inhibits the growth of a variety of human tumor cell lines in vitro and selectively induces apoptosis in cancer cell lines but not in normal cells. purity: 99%
Catalog No. 50-258-0383 Supplier TARGETMOL CHEMICALS INC Supplier No. T310205MG
Add to Cart
Add to Cart
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. CP 461 (UNII-68OJX9I7DT) a specific PDE2A inhibitor is a novel pro-apoptotic compound that inhibits cyclic GMP phosphodiesterase but not cyclooxygenase-1 or -2. CP 461 inhibits the growth of a variety of human tumor cell lines in vitro and selectively induces apoptosis in cancer cell lines but not in normal cells. purity: 99%