Learn More
TARGETMOL CHEMICALS INC CP 465022 5MG
SDP
Catalog No. 502581021
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CP 465022 hydrochloride is a potent and selective noncompetitive antagonist of AMPA receptor with anticonvulsant activity. CP 465022 hydrochloride inhibits Kainate-induced response with an IC50 of 25 nM in rat cortical neurons. purity: 98%
Catalog No. 50-258-1021 Supplier TARGETMOL CHEMICALS INC Supplier No. T215845MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CP 465022 hydrochloride is a potent and selective noncompetitive antagonist of AMPA receptor with anticonvulsant activity. CP 465022 hydrochloride inhibits Kainate-induced response with an IC50 of 25 nM in rat cortical neurons. purity: 98%