Learn More
TARGETMOL CHEMICALS INC CP-466722 10MG
SDP
Catalog No. 502576552
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CP-466722 an effective and reversible ATM inhibitor does not inhibit ATR and PI3K or PIKK family members in cells. purity: 99%
Catalog No. 50-257-6552 Supplier TARGETMOL CHEMICALS INC Supplier No. T626110MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CP-466722 an effective and reversible ATM inhibitor does not inhibit ATR and PI3K or PIKK family members in cells. purity: 99%