Learn More
Selleck Chemical LLC CP-466722 2mg 1080622-86-1
SDP
Catalog No. 508615
Encompass_Preferred
CP-466722 is a potent and reversible ATM inhibitor, does not affect ATR and inhibits PI3K or PIKK family members in cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-861-5 Supplier Selleck Chemical LLC Supplier No. S22452MG
CP-466722 is a potent and reversible ATM inhibitor, does not affect ATR and inhibits PI3K or PIKK family members in cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.