Learn More
Guardian™ CP-5, Chemical Resistant Smooth Finish 5 mil Butyl Gloves
SDP
Catalog No. 19321188
Click to view available options
Size:
Large
Medium
Small
X-Large
X-Small
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-321-188 X-Large
19-321-184 X-Small
19-321-185 Small
19-321-186 Medium
19-321-187 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-321-188 Supplier Guardian™ Supplier No. 50505
This item is not returnable. View return policy
USA made, 14 in. butyl gloves offer protection against a variety of hazardous chemicals.
  • Butyl material resists oxidation, ozone corrosion and abrasions
  • Reusable and easy to clean
  • Fitted hand style with smooth texture
  • Protects against wide range of hazardous chemicals
  • 5 mil thickness, 14 in. length
  • Made in the USA
Specifications
Color Black
Hand Specific
Latex Free Yes
Material Butyl
Powdered Powdered
Size X-Large
Thickness (English) 5 mil
Thickness (Metric) 0.12 mm
Glove Cuff Style Gauntlet
Grip Design Smooth
Lining Unlined
Model 50505
Length (English) 14 in.
Length (Metric) 35.5 cm