Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cp-506 mesylate | 2227304-19-8 | ≥98.0% | 408.40 | C17H17FN4O4S | 10 MG
SDP

Catalog No. 5000285832
Encompass_Preferred

CP-506 mesylate is the mesylate salt of CP-506, a small-molecule anticancer compound reported as a selective MAP4K2 inhibitor with potent in vitro activity and demonstrated in vivo pharmacokinetic properties. It is supplied for laboratory research use and is provided with purity and storage specifications suitable for preclinical studies.

  • Potent MAP4K2 inhibitor with IC50 ≈ 6.2 nM.
  • Mesylate salt form for improved handling and stability.
  • Molecular weight 408.40 and formula C17H17FN4O4S.
  • High purity (≥98.0%).
  • DMSO solubility approximately 100 mg/mL.
  • Storage: 8°C, protect from light; in solvent freeze at -80°C for long term.

Catalog No. 50-002-85832 Supplier Medchemexpress LLC Supplier No. HY149769A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CP-506 mesylate is the mesylate salt of CP-506, a small-molecule anticancer compound reported as a selective MAP4K2 inhibitor with potent in vitro activity and demonstrated in vivo pharmacokinetic properties. It is supplied for laboratory research use and is provided with purity and storage specifications suitable for preclinical studies.

  • Potent MAP4K2 inhibitor with IC50 ≈ 6.2 nM.
  • Mesylate salt form for improved handling and stability.
  • Molecular weight 408.40 and formula C17H17FN4O4S.
  • High purity (≥98.0%).
  • DMSO solubility approximately 100 mg/mL.
  • Storage: 8°C, protect from light; in solvent freeze at -80°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.