Learn More
Medchemexpress LLC HY-13302 5mg Medchemexpress, CP-547632 CAS:252003-65-9 Purity:>98%
SDP
Catalog No. 501932660
Encompass_Preferred
Medchemexpress, HY-13302 5mg CP-547632 CAS:252003-65-9 CP-547632 is a well-tolerated, orally-bioavailable inhibitor of the VEGFR-2 and basic fibroblast growth factor ( FGF ) kinases with IC 50 s of 11 nM and 9 nM, respectively. CP-547632 is an ATP-competitive kinase inhibitor and it is selective relative to epidermal growth factor receptor, platelet-derived growth factor β, and other related TKs. CP-547632 has antitumor efficacy [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2660 Supplier Medchemexpress LLC Supplier No. HY133025MG
Medchemexpress, HY-13302 5mg CP-547632 CAS:252003-65-9 CP-547632 is a well-tolerated, orally-bioavailable inhibitor of the VEGFR-2 and basic fibroblast growth factor ( FGF ) kinases with IC 50 s of 11 nM and 9 nM, respectively. CP-547632 is an ATP-competitive kinase inhibitor and it is selective relative to epidermal growth factor receptor, platelet-derived growth factor β, and other related TKs. CP-547632 has antitumor efficacy [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.