Learn More
Medchemexpress LLC HY-13302B 5mg Medchemexpress, CP-547632 (hydrochloride) CAS:252003-71-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13302B5MG
Encompass_Preferred
Medchemexpress, HY-13302B 5mg CP-547632 (hydrochloride) CAS:252003-71-7 CP-547632 hydrochloride is a well-tolerated, orally-bioavailable inhibitor of the VEGFR-2 and basic fibroblast growth factor ( FGF ) kinases with IC 50 s of 11 nM and 9 nM, respectively. CP-547632 hydrochloride is an ATP-competitive kinase inhibitor and it is selective relative to epidermal growth factor receptor, platelet-derived growth factor β, and other related TKs. CP-547632 hydrochloride has antitumor efficacy [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2661