Learn More
Medchemexpress LLC CP-547632 hydrochloride | 252003-71-7 | MFCD20268381 | 99.9% | C20H25BrClF2N5O3S | 10MG
SDP
Catalog No. 5000258779
Encompass_Preferred
CP-547632 hydrochloride is the hydrochloride salt form of a potent, orally active ATP-competitive inhibitor of VEGFR-2 and FGF kinases, with reported IC50 values of 11 nM and 9 nM, respectively. It demonstrates selectivity versus EGFR and PDGFRβ and has shown antitumor efficacy in preclinical models.
  • ATP-competitive VEGFR-2 and FGF kinase inhibition.
  • Low-nanomolar potency (IC50s ≈11 nM and ≈9 nM).
  • Selective profile versus related tyrosine kinases.
  • Suitable for in vitro and in vivo preclinical studies.
  • High supplied purity (~99.9%).
Catalog No. 50-002-58779 Supplier Medchemexpress LLC Supplier No. HY13302B10MG
CP-547632 hydrochloride is the hydrochloride salt form of a potent, orally active ATP-competitive inhibitor of VEGFR-2 and FGF kinases, with reported IC50 values of 11 nM and 9 nM, respectively. It demonstrates selectivity versus EGFR and PDGFRβ and has shown antitumor efficacy in preclinical models.
  • ATP-competitive VEGFR-2 and FGF kinase inhibition.
  • Low-nanomolar potency (IC50s ≈11 nM and ≈9 nM).
  • Selective profile versus related tyrosine kinases.
  • Suitable for in vitro and in vivo preclinical studies.
  • High supplied purity (~99.9%).