Learn More
Enzo Life Sciences CP-55,940 (25mg). CAS: 83002-04-4
SDP
Supplier:  Enzo Life Sciences BMLCR1020025
Encompass_Preferred
Cannabinoid CB1 and CB2 agonist which is more potent than δ9-THC. Kd’s for cloned/transfected CB1 and CB2: 2.6 & 3.7 nM. Has been reported to induce the immediate early gene Krox-24 in corpus striatum and cultured astrocytes. Stimulates proliferation of stimulated B-cells at low nM concentrations. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (4mg/ml) or ethanol. Long Term Storage: -20°C.
Catalog No. 50-201-0466