Learn More
Eurofins Discoverx CP 55,940, 5 mg. (92-1343)
Supplier:  Eurofins Discoverx 921343
Encompass
CP 55,940 is a synthetic cannabinoid molecule developed by Pfizer that has similar but more potent effects to THC in behavioral and receptor binding tests. It is a cannabinoid receptor agonist, with EC50=0.2 and 0.3 nM for CB1 and CB2 respectively. CP 55,490 is reported as both an agonist (EC50 = 5 nM) and antagonist for GRP55 depending on the assay and dosages uses. Currently used to study the endocannabinoid system. Provided as 5mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9852