Learn More
Cayman Chemical CP 55 940d11 GC MS Standards
SDP
Supplier:  Cayman Chemical 10703100 UG
Encompass_Preferred
An internal standard for the quantification of (±)-CP 55,940 by GC- or LC-MS; intended for research and forensic applications
Catalog No. 50-288-9605