Learn More
Cayman Chemical CP 55 940d11 GC MS Standards
SDP
Catalog No. 502890185
Encompass_Preferred
An internal standard for the quantification of (±)-CP 55,940 by GC- or LC-MS; intended for research and forensic applications
Catalog No. 50-289-0185 Supplier Cayman Chemical Supplier No. 10703500 UG
An internal standard for the quantification of (±)-CP 55,940 by GC- or LC-MS; intended for research and forensic applications